siRNA Details
| Gene Symbol: THUMPD3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027007.3 | 3 | 733 | |||
| NR_132780.1 | 4 | 807 | ||||
| GenBank mRNA | AK093640.1 | - | 2 | 1858 | ||
| AL110136.1 | - | 3 | 729 | |||
| BC044625.2 | - | 1 | 1844 | |||
| BC052961.1 | - | 3 | 3105 | |||
| DB567546.1 | - | 1 | 400 | |||

THUMPD3-AS1
Genomic Map