siRNA Details
| Gene Symbol: FAM106CP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026810.1 | 1 | 91 | |||
| GenBank mRNA | DN989334.1 | - | 1 | 100 | ||
| Gene Symbol: FAM106B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126160.1 | 1 | 91 | |||

FAM106CP
Genomic Map