siRNA Details
| Gene Symbol: LOC645752 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027024.1 | 19 | 2506 | |||
| GenBank mRNA | AK294847.1 | - | 3 | 801 | ||
| AK308609.1 | - | 4 | 842 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC645752| Gene Symbol: LOC645752 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027024.1 | 19 | 2506 | |||
| GenBank mRNA | AK294847.1 | - | 3 | 801 | ||
| AK308609.1 | - | 4 | 842 | |||