siRNA Details
| Gene Symbol: GGA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291641.1 | NP_001278570.1 | 2 | 165 | ||
| XM_017024387.1 | XP_016879876.1 | 2 | 119 | |||
| XM_017024388.1 | XP_016879877.1 | 2 | 131 | |||
| XM_017024392.1 | XP_016879881.1 | 2 | 121 | |||
| GenBank mRNA | AL833603.1 | - | 2 | 105 | ||
| HY030361.1 | - | 2 | 164 | |||

GGA3
Genomic Map