siRNA Details
| Gene Symbol: C20orf24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026562.3 | 2, 3 | 435 | |||
| GenBank mRNA | BC004446.1 | - | 2, 3 | 360 | 2819807 | |
| BQ086248.1 | - | 2, 3 | 384 | |||
| BU174619.1 | - | 403 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C20orf24| Gene Symbol: C20orf24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026562.3 | 2, 3 | 435 | |||
| GenBank mRNA | BC004446.1 | - | 2, 3 | 360 | 2819807 | |
| BQ086248.1 | - | 2, 3 | 384 | |||
| BU174619.1 | - | 403 | ||||