siRNA Details
| Gene Symbol: NXF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028089.1 | 16 | 1499 | |||
| GenBank mRNA | AJ277654.1 | - | 16 | 1499 | ||
| AJ277655.1 | - | 16 | 1474 | |||
| AJ277656.1 | - | 16 | 1410 | |||
| AJ277657.1 | - | 14 | 1285 | |||
| AJ277658.1 | - | 14 | 1404 | |||

NXF5
Genomic Map