siRNA Details
Gene Symbol: ADGRE4P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024075.1 | 4 | 392 | |||
GenBank mRNA | AF489700.1 | - | 4 | 392 | ||
AY181245.1 | - | 3 | 296 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ADGRE4P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024075.1 | 4 | 392 | |||
GenBank mRNA | AF489700.1 | - | 4 | 392 | ||
AY181245.1 | - | 3 | 296 |