siRNA Details
| Gene Symbol: POPDC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024539.1 | 1, 2 | 59 | |||
| XM_011536067.2 | XP_011534369.1 | 1, 2 | 307 | |||
| GenBank mRNA | AK225989.1 | - | 1, 2 | 59 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BP369262.1 | - | 1, 2 | 59 | Chr. 6 - 105158709 - 105176255 [-] on Build GRCh38 | |

POPDC3
Genomic Map