siRNA Details
| Gene Symbol: GUSBP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003675.2 | 1 | 1090 | |||
| GenBank mRNA | BC025996.2 | - | 1 | 1090 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GUSBP5| Gene Symbol: GUSBP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003675.2 | 1 | 1090 | |||
| GenBank mRNA | BC025996.2 | - | 1 | 1090 | ||