siRNA Details
| Gene Symbol: SGK494 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001174103.1 | NP_001167574.1 | 11, 12 | 1039 | ||
| GenBank mRNA | BC147005.1 | - | 2, 3 | 160 | ||
| BC147009.1 | - | 2, 3 | 160 | |||
| EB388892.1 | - | 2, 3 | 215 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SGK494| Gene Symbol: SGK494 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001174103.1 | NP_001167574.1 | 11, 12 | 1039 | ||
| GenBank mRNA | BC147005.1 | - | 2, 3 | 160 | ||
| BC147009.1 | - | 2, 3 | 160 | |||
| EB388892.1 | - | 2, 3 | 215 | |||