siRNA Details
| Gene Symbol: SGK494 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001174103.1 | NP_001167574.1 | 12 | 1245 | ||
| GenBank mRNA | BC001565.1 | - | 1 | 441 | ||
| BC147005.1 | - | 3 | 366 | |||
| BC147009.1 | - | 3 | 366 | |||
| EB388892.1 | - | 3 | 421 | |||

SGK494
Genomic Map