siRNA Details
Gene Symbol: C7orf13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026865.1 | 1 | 2052 | |||
GenBank mRNA | AF063598.1 | - | 1 | 1819 | ||
AF439977.1 | - | 1 | 2052 | |||
AK056371.1 | - | 1 | 1968 | |||
BC031037.1 | - | 1 | 1937 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA335849.1 | - | 1 | 365 | Chr. 7 - 156638375 - 156638967 [-] on Build GRCh38 |
