siRNA Details
| Gene Symbol: CD27-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015382.2 | 6 | 2082 | |||
| GenBank mRNA | AK056500.1 | - | 6 | 2082 | ||
| AK128574.1 | - | 2 | 2954 | |||
| BC033973.1 | - | 7 | 1055 | |||
| BC041960.1 | - | 3 | 1378 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CD27-AS1| Gene Symbol: CD27-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015382.2 | 6 | 2082 | |||
| GenBank mRNA | AK056500.1 | - | 6 | 2082 | ||
| AK128574.1 | - | 2 | 2954 | |||
| BC033973.1 | - | 7 | 1055 | |||
| BC041960.1 | - | 3 | 1378 | |||