siRNA Details
| Gene Symbol: LOC100129550 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024618.1 | 1 | 2722 | |||
| GenBank mRNA | AK127315.1 | - | 1 | 2296 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100129550| Gene Symbol: LOC100129550 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024618.1 | 1 | 2722 | |||
| GenBank mRNA | AK127315.1 | - | 1 | 2296 | ||