siRNA Details
| Gene Symbol: RPL34-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026968.1 | 4 | 2907 | |||
| GenBank mRNA | AK094992.1 | - | 4 | 2850 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RPL34-AS1| Gene Symbol: RPL34-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026968.1 | 4 | 2907 | |||
| GenBank mRNA | AK094992.1 | - | 4 | 2850 | ||