siRNA Details
| Gene Symbol: PCDHGB8P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001297.1 | 1 | 909 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AF152529.3 | - | 1 | 909 | Chr. 5 - 141426286 - 141428258 [+] on Build GRCh38 | |

PCDHGB8P
Genomic Map