siRNA Details
| Gene Symbol: LINC01098 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028342.1 | 6 | 808 | |||
| GenBank mRNA | BC033326.1 | - | 6 | 808 | ||
| BC113482.1 | - | 4 | 373 | |||
| BC113484.1 | - | 4 | 373 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01098| Gene Symbol: LINC01098 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028342.1 | 6 | 808 | |||
| GenBank mRNA | BC033326.1 | - | 6 | 808 | ||
| BC113482.1 | - | 4 | 373 | |||
| BC113484.1 | - | 4 | 373 | |||