siRNA Details
| Gene Symbol: USP32P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003190.2 | 3, 4 | 660 | |||
| GenBank mRNA | AK311696.1 | - | 4, 5 | 862 | ||
| BC039345.1 | - | 2, 3 | 324 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

USP32P1| Gene Symbol: USP32P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003190.2 | 3, 4 | 660 | |||
| GenBank mRNA | AK311696.1 | - | 4, 5 | 862 | ||
| BC039345.1 | - | 2, 3 | 324 | |||