siRNA Details
| Gene Symbol: USP32P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003190.2 | 6 | 1546 | |||
| GenBank mRNA | AK302945.1 | - | 2 | 651 | ||
| AK316466.1 | - | 4 | 1264 | |||
| AY363305.1 | - | 2 | 928 | |||
| BC039345.1 | - | 6 | 1303 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

USP32P1| Gene Symbol: USP32P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003190.2 | 6 | 1546 | |||
| GenBank mRNA | AK302945.1 | - | 2 | 651 | ||
| AK316466.1 | - | 4 | 1264 | |||
| AY363305.1 | - | 2 | 928 | |||
| BC039345.1 | - | 6 | 1303 | |||