siRNA Details
| Gene Symbol: LOC154449 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002787.2 | 3 | 456 | |||
| GenBank mRNA | AK058183.1 | - | 3 | 456 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC154449| Gene Symbol: LOC154449 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002787.2 | 3 | 456 | |||
| GenBank mRNA | AK058183.1 | - | 3 | 456 | ||