siRNA Details
| Gene Symbol: NDUFB2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024454.1 | 2 | 1031 | |||
| GenBank mRNA | AK129584.1 | - | 2 | 1031 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NDUFB2-AS1| Gene Symbol: NDUFB2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024454.1 | 2 | 1031 | |||
| GenBank mRNA | AK129584.1 | - | 2 | 1031 | ||