siRNA Details
Gene Symbol: LINC00602 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027284.1 | 1 | 841 | |||
GenBank mRNA | AK094116.1 | - | 1 | 841 | ||
BC110806.1 | - | 1 | 765 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00602 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027284.1 | 1 | 841 | |||
GenBank mRNA | AK094116.1 | - | 1 | 841 | ||
BC110806.1 | - | 1 | 765 |