siRNA Details
| Gene Symbol: LINC00602 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027284.1 | 1 | 121 | |||
| GenBank mRNA | AK094116.1 | - | 1 | 121 | ||
| BC110806.1 | - | 1 | 45 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00602| Gene Symbol: LINC00602 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027284.1 | 1 | 121 | |||
| GenBank mRNA | AK094116.1 | - | 1 | 121 | ||
| BC110806.1 | - | 1 | 45 | |||