siRNA Details
| Gene Symbol: ZNF519 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033354.1 | 2 | 302 | |||
| XM_017025562.1 | XP_016881051.1 | 4 | 9964 | |||
| XM_017025563.1 | XP_016881052.1 | 5 | 5964 | |||
| XM_017025564.1 | XP_016881053.1 | 4 | 5703 | |||
| XM_017025565.1 | XP_016881054.1 | 4 | 5703 | |||
| GenBank mRNA | AK096748.1 | - | 3 | 368 | ||
| BC037787.1 | - | 2 | 270 | |||
| DA525188.1 | - | 2 | 302 | |||

ZNF519
Genomic Map