siRNA Details
| Gene Symbol: LOC100129215 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AW504969.1 | - | 1 | 122 | ||
| Gene Symbol: KLHDC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK001742.1 | - | 4 | 1682 | ||
| AK024496.1 | - | 1 | 3204 | |||
| AY251274.1 | - | 1 | 3291 | |||

LOC100129215
Genomic Map