siRNA Details
| Gene Symbol: KLHDC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006721203.2 | XP_006721266.1 | 12 | 2075 | ||
| XM_017023344.1 | XP_016878833.1 | 12 | 2060 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KLHDC4| Gene Symbol: KLHDC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006721203.2 | XP_006721266.1 | 12 | 2075 | ||
| XM_017023344.1 | XP_016878833.1 | 12 | 2060 | |||