siRNA Details
| Gene Symbol: DICER1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015415.1 | 4 | 484 | |||
| GenBank mRNA | AK127179.1 | - | 2 | 2014 | ||
| BI754942.1 | - | 4 | 410 | |||
| BX648898.1 | - | 1 | 5860 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BE788052.1 | - | 499 | Chr. - - [-] on Build | ||

DICER1-AS1
Genomic Map