siRNA Details
Gene Symbol: SSTR5-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027242.1 | 1 | 42 | |||
GenBank mRNA | BC084558.1 | - | 1 | 42 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SSTR5-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027242.1 | 1 | 42 | |||
GenBank mRNA | BC084558.1 | - | 1 | 42 |