siRNA Details
| Gene Symbol: SSTR5-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027242.1 | 4 | 1944 | |||
| GenBank mRNA | AK056814.1 | - | 4 | 1888 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000569832.5 | - | 4 | 1886 | Chr. 16 - 1064093 - 1078673 [-] on Build GRCh38 | |

SSTR5-AS1
Genomic Map