siRNA Details
| Gene Symbol: SSTR5-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027242.1 | 4 | 1664 | |||
| GenBank mRNA | AK056814.1 | - | 4 | 1608 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SSTR5-AS1| Gene Symbol: SSTR5-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027242.1 | 4 | 1664 | |||
| GenBank mRNA | AK056814.1 | - | 4 | 1608 | ||