siRNA Details
| Gene Symbol: LRTOMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026886.3 | 3, 4 | 771 | |||
| GenBank mRNA | AK311690.1 | - | 3, 4 | 241 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DB201990.1 | - | 3, 4 | 241 | Chr. 11 - 72080861 - 72093656 [+] on Build GRCh38 | |

LRTOMT
Genomic Map