siRNA Details
Gene Symbol: LRTOMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026886.3 | 4 | 933 | |||
GenBank mRNA | AK311690.1 | - | 4 | 403 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB201990.1 | - | 4 | 403 | Chr. 11 - 72080861 - 72093656 [+] on Build GRCh38 |
