siRNA Details
| Gene Symbol: TTC31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027749.1 | 2, 3 | 151 | |||
| XM_011533040.1 | XP_011531342.1 | 2, 3 | 192 | |||
| XM_017004708.1 | XP_016860197.1 | 2, 3 | 148 | |||
| GenBank mRNA | AK303105.1 | - | 2, 3 | 137 | ||
| BC045536.1 | - | 2, 3 | 142 | |||
| BC099729.1 | - | 2, 3 | 137 | |||
| BX648131.1 | - | 2, 3 | 136 | |||

TTC31
Genomic Map