siRNA Details
Gene Symbol: DTX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017011728.1 | XP_016867217.1 | 6, 7 | 1476 |
Gene Symbol: DTX2P1-UPK3BP1-PMS2P11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023383.1 | 4, 5 | 777 | |||
GenBank mRNA | BM709868.1 | - | 1, 2 | 92 | ||
U38980.1 | - | 4, 5 | 662 |
