siRNA Details
| Gene Symbol: LINC00244 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024119.1 | 1 | 145 | |||
| GenBank mRNA | AF107456.1 | - | 1 | 145 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00244| Gene Symbol: LINC00244 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024119.1 | 1 | 145 | |||
| GenBank mRNA | AF107456.1 | - | 1 | 145 | ||