siRNA Details
Gene Symbol: ARRDC3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027435.1 | 1, 2 | 578 | |||
GenBank mRNA | BC043415.2 | - | 1, 2 | 567 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ARRDC3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027435.1 | 1, 2 | 578 | |||
GenBank mRNA | BC043415.2 | - | 1, 2 | 567 |