siRNA Details
| Gene Symbol: LINC01126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027251.1 | 1 | 1055 | |||
| GenBank mRNA | BC032412.1 | - | 1 | 925 | 5216225 | |
| CR738297.1 | - | 1 | 444 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01126| Gene Symbol: LINC01126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027251.1 | 1 | 1055 | |||
| GenBank mRNA | BC032412.1 | - | 1 | 925 | 5216225 | |
| CR738297.1 | - | 1 | 444 | |||