siRNA Details
| Gene Symbol: ZNF525 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003699.1 | 4 | 5449 | |||
| GenBank mRNA | AB075859.1 | - | 1 | 4400 | ||
| AK097037.1 | - | 4 | 1221 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF525| Gene Symbol: ZNF525 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003699.1 | 4 | 5449 | |||
| GenBank mRNA | AB075859.1 | - | 1 | 4400 | ||
| AK097037.1 | - | 4 | 1221 | |||