siRNA Details
| Gene Symbol: TPTE2P3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002793.1 | 24 | 2440 | |||
| Gene Symbol: TPTE2P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC041842.1 | - | 7 | 3660 | ||

TPTE2P3
Genomic Map