siRNA Details
| Gene Symbol: TPTE2P3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002793.1 | 5, 6 | 819 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TPTE2P3| Gene Symbol: TPTE2P3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002793.1 | 5, 6 | 819 | |||