siRNA Details
| Gene Symbol: DEFA10P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029386.1 | 2 | 228 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AY746435.1 | - | 2 | 228 | Chr. 8 - 6968141 - 6969113 [-] on Build GRCh38 | |

DEFA10P
Genomic Map