siRNA Details
| Gene Symbol: LOC728024 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003671.2 | 1 | 129 | |||
| GenBank mRNA | BI758198.1 | - | 1 | 133 | ||
| CR977513.1 | - | 1 | 36 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC728024| Gene Symbol: LOC728024 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003671.2 | 1 | 129 | |||
| GenBank mRNA | BI758198.1 | - | 1 | 133 | ||
| CR977513.1 | - | 1 | 36 | |||