siRNA Details
| Gene Symbol: HCG4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001317.3 | 1 | 2400 | |||
| GenBank mRNA | AK056727.1 | - | 2 | 2361 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AF036973.1 | - | 2 | 1228 | Chr. 6 - 29924695 - 29926086 [-] on Build GRCh38 | |

HCG4B
Genomic Map