siRNA Details
| Gene Symbol: SSR4P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027292.2 | 1 | 1768 | |||
| GenBank mRNA | BC004343.1 | - | 1 | 1768 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SSR4P1| Gene Symbol: SSR4P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027292.2 | 1 | 1768 | |||
| GenBank mRNA | BC004343.1 | - | 1 | 1768 | ||