siRNA Details
Gene Symbol: SSR4P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027292.2 | 1 | 1602 | |||
GenBank mRNA | BC004343.1 | - | 1 | 1602 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SSR4P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027292.2 | 1 | 1602 | |||
GenBank mRNA | BC004343.1 | - | 1 | 1602 |