siRNA Details
| Gene Symbol: SLC25A21-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033240.1 | 1 | 1240 | |||
| GenBank mRNA | AK057476.1 | - | 1 | 1163 | ||
| BC040167.1 | - | 1 | 1158 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC25A21-AS1| Gene Symbol: SLC25A21-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033240.1 | 1 | 1240 | |||
| GenBank mRNA | AK057476.1 | - | 1 | 1163 | ||
| BC040167.1 | - | 1 | 1158 | |||