siRNA Details
Gene Symbol: SLC25A21-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033240.1 | 1 | 792 | |||
GenBank mRNA | AK057476.1 | - | 1 | 715 | ||
BC012470.1 | - | 1 | 792 | |||
BC040167.1 | - | 1 | 710 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SLC25A21-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033240.1 | 1 | 792 | |||
GenBank mRNA | AK057476.1 | - | 1 | 715 | ||
BC012470.1 | - | 1 | 792 | |||
BC040167.1 | - | 1 | 710 |