siRNA Details
| Gene Symbol: FAM197Y2P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001553.1 | 3 | 383 | |||
| GenBank mRNA | AK129785.1 | - | 2 | 187 | ||
| Gene Symbol: FAM197Y5P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046300.1 | 3 | 383 | |||

FAM197Y2P
Genomic Map