siRNA Details
| Gene Symbol: TTTY19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001549.1 | 1, 2 | 94 | |||
| GenBank mRNA | AF527831.1 | - | 1, 2 | 94 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TTTY19| Gene Symbol: TTTY19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001549.1 | 1, 2 | 94 | |||
| GenBank mRNA | AF527831.1 | - | 1, 2 | 94 | ||