siRNA Details
| Gene Symbol: TTTY19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001549.1 | 1 | 21 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AF527831.3 | - | 1 | 21 | Chr. Y - 8704472 - 8705283 [+] on Build GRCh38 | |

TTTY19
Genomic Map